missing translation for 'onlineSavingsMsg'
Learn More

ITE, TRC

Product Code. 30400274
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30400274 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30400274 Supplier Toronto Research Chemicals Supplier No. TRCI9420305MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material ITE
CAS 448906-42-1
Molecular Formula C14H10N2O3S
Quantity 5 mg
Synonym 2-(1H-Indol-3-ylcarbonyl)-4-thiazolecarboxylic Acid Methyl Ester, 2-(1'H-Indole-3'-carbonyl)thiazole-4-carboxylic Acid Methyl Ester
InChI Formula InChI=1S/C14H10N2O3S/c1-19-14(18)11-7-20-13(16-11)12(17)9-6-15-10-5-3-2-4-8(9)10/h2-7,15H,1H3
SMILES COC(=O)c1csc(n1)C(=O)c2c[nH]c3ccccc23
IUPAC Name methyl 2-(1H-indole-3-carbonyl)-1,3-thiazole-4-carboxylate
Molecular Weight (g/mol) 286.31
Formula Weight 286.0412
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.