missing translation for 'onlineSavingsMsg'
Learn More

ITE, MedChemExpress

Product Code. 16487718
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16487718 10 mg 10mg
16433697 50 mg 50mg
16420188 5 mg 5mg
16447586 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16487718 Supplier Medchem Express Supplier No. HY19317
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress ITE is a potent endogenous agonist of aryl hydrocarbon receptor (AhR), binding directly to AHR, with a Ki of 3 nM. ITE also has immunosuppressive activity.

ITE is a potent endogenous agonist of aryl hydrocarbon receptor (AhR), binding directly to AHR, with a Ki of 3 nM. ITE also has immunosuppressive activity.

Specifications

Chemical Name or Material ITE
CAS 448906-42-1
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C14H10N2O3S
Quantity 10 mg
Solubility Information DMSO : ≥ 41 mg/mL (143.20 mM)
SMILES O=C(C1=CSC(C(C2=CNC3=C2C=CC=C3)=O)=N1)OC
Molecular Weight (g/mol) 286.31
Formula Weight 286.31
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.