missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ ITD 1
Description
ITD 1 is a selective inhibitor of TGF-β signaling (IC50 = 0.85 μM); displays little or no inhibition of activin, Wnt or BMP signaling pathways. Induces proteasomal degradation of the TGF-β type II receptor. Inhibits TGF-β-induced mesoderm formation from mouse embryonic stem cells (ESCs) during early differentiation; selectively promotes the differentiation of ESCs to cardiomyocytes in vitro between days 3-5. Does not induce differentiation of vascular smooth muscle cells or endothelial cells.
Specifications
Specifications
| Purity | >98% |
| Content And Storage | -20°C |
| Quantity | 50 mg |
| Product Type | ITD 1 |
| Molecular Weight (g/mol) | 415.52 |
| Product Line | Tocris |
| Inhibitors | ITD 1 |
For Research Use Only
Produkttitel
Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.
Hittar du en möjlighet till förbättring?