missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ ITD 1

Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
ITD 1 is a selective inhibitor of TGF-β signaling (IC50 = 0.85 μM); displays little or no inhibition of activin, Wnt or BMP signaling pathways. Induces proteasomal degradation of the TGF-β type II receptor. Inhibits TGF-β-induced mesoderm formation from mouse embryonic stem cells (ESCs) during early differentiation; selectively promotes the differentiation of ESCs to cardiomyocytes in vitro between days 3-5. Does not induce differentiation of vascular smooth muscle cells or endothelial cells.
Specifications
Specifications
| Purity | >98% |
| Content And Storage | -20°C |
| Quantity | 10 mg |
| Product Type | ITD 1 |
| Molecular Weight (g/mol) | 415.52 |
| Inhibitors | ITD 1 |
For Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction