missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ITCH Recombinant Rabbit Monoclonal Antibody (23GB6255), Invitrogen™

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA553006
This item is not returnable.
View return policy
Description
Atrophin-1 contains a polyglutamine repeat, expansion of which is responsible for dentatorubral and pallidoluysian atrophy. The protein encoded by this gene interacts with atrophin-1. This encoded protein is a closely related member of the NEDD4-like protein family. This family of proteins are E3 ubiquitin-ligase molecules and regulate key trafficking decisions, including targeting of proteins to proteosomes or lysosomes. This encoded protein contains four tandem WW domains and a HECT domain. It can act as a transcriptional corepressor of p45/NFE2 and may participate in the regulation of immune responses by modifying Notch-mediated signaling. It is highly similar to the mouse Itch protein, which has been implicated in the regulation and differentiation of erythroid and lymphoid cells.
Specifications
| ITCH | |
| Recombinant Monoclonal | |
| Unconjugated | |
| ITCH | |
| 6720481N21Rik; 8030492O04Rik; A130065M08; ADMFD; AIF4; AIP4; atrophin-1 interacting protein 4; Atrophin-1-interacting protein 4; C230047C07Rik; dJ468O1.1; E3 ubiquitin-protein ligase Itchy; E3 ubiquitin-protein ligase Itchy homolog; HECT-type E3 ubiquitin transferase Itchy homolog; ITCH; itch E3 ubiquitin ligase; itchy E3 ubiquitin protein ligase; itchy E3 ubiquitin protein ligase homolog; itchy, E3 ubiquitin protein ligase; NAPP1; NFE2-associated polypeptide 1 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 16396, 311567, 83737 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 23GB6255 | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q8C863, Q96J02 | |
| ITCH | |
| A synthesized peptide derived from human ITCH (10-66AA). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction