missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ITCH/AIP4 Rabbit anti-Human, Mouse, Rat, Clone: 7I7B10, Novus Biologicals™
Rabbit Monoclonal Antibody
£150.00 - £425.00
Specifications
| Antigen | ITCH/AIP4 |
|---|---|
| Clone | 7I7B10 |
| Dilution | Western Blot 1:500 - 1:2000, Knockout Validated |
| Applications | Western Blot, KnockDown |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18345034
|
Novus Biologicals
NBP3-16669-20UL |
20 μg |
£150.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18341805
|
Novus Biologicals
NBP3-16669-100UL |
100 μg |
£425.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
ITCH/AIP4 Monoclonal antibody specifically detects ITCH/AIP4 in Human, Mouse, Rat samples. It is validated for Western Blot, KnockDownSpecifications
| ITCH/AIP4 | |
| Western Blot 1:500 - 1:2000, Knockout Validated | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| AIF4, AIP4dJ468O1.1 (atrophin 1 interacting protein 4 (AIP4)), atrophin-1 interacting protein 4, Atrophin-1-interacting protein 4, dJ468O1.1, EC 6.3.2, EC 6.3.2.-, Itch, itchy (mouse homolog) E3 ubiquitin protein ligase, itchy E3 ubiquitin protein ligase homolog (mouse), itchy homolog E3 ubiquitin protein ligase, NAPP1E3 ubiquitin-protein ligase Itchy homolog, NFE2-associated polypeptide 1, ubiquitin protein ligase ITCH | |
| A synthetic peptide corresponding to a sequence within amino acids 1-100 of human ITCH/AIP4 (Q96J02). MSDSGSQLGSMGSLTMKSQLQITVISAKLKENKKNWFGPSPYVEVTVDGQSKKTEKCNNTNSPKWKQPLTVIVTPVSKLHFRVWSHQTLKSDVLLGTAAL | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
| 7I7B10 | |
| Western Blot, KnockDown | |
| Unconjugated | |
| Rabbit | |
| Apoptosis, Cell Biology, Immunology, Signal Transduction | |
| PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
| 83737 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title