missing translation for 'onlineSavingsMsg'
Learn More

ITCH/AIP4 Antibody, Novus Biologicals™

Product Code. 18286380 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18286380 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18286380 Supplier Novus Biologicals Supplier No. NBP118881

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

ITCH/AIP4 Polyclonal specifically detects ITCH/AIP4 in Human samples. It is validated for Western Blot, Immunoprecipitation (Negative).
TRUSTED_SUSTAINABILITY

Specifications

Antigen ITCH/AIP4
Applications Western Blot, Immunoprecipitation
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:100-1:2000, Immunoprecipitation
Formulation TBS and 0.1% BSA with 0.09% Sodium Azide
Gene Accession No. AAH11571.1
Gene Alias AIF4, AIP4dJ468O1.1 (atrophin 1 interacting protein 4 (AIP4)), atrophin-1 interacting protein 4, Atrophin-1-interacting protein 4, dJ468O1.1, EC 6.3.2, EC 6.3.2.-, Itch, itchy (mouse homolog) E3 ubiquitin protein ligase, itchy E3 ubiquitin protein ligase homolog (mouse), itchy homolog E3 ubiquitin protein ligase, NAPP1E3 ubiquitin-protein ligase Itchy homolog, NFE2-associated polypeptide 1, ubiquitin protein ligase ITCH
Gene Symbols ITCH
Host Species Rabbit
Immunogen The immunogen recognized by this antibody maps to a region between residue 650 to 700 of human itchy homolog E3 ubiquitin protein ligase using the numbering given in entry AAH11571.1 (GeneID 83737).
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 83737
Target Species Human
Content And Storage Store at 4C. Do not freeze.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.