missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ITCH/AIP4 Antibody, mFluor Violet 500 SE, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-81837MFV500
This item is not returnable.
View return policy
Description
ITCH/AIP4 Polyclonal antibody specifically detects ITCH/AIP4 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA
Specifications
| ITCH/AIP4 | |
| Polyclonal | |
| Western Blot, ELISA | |
| AIF4, AIP4dJ468O1.1 (atrophin 1 interacting protein 4 (AIP4)), atrophin-1 interacting protein 4, Atrophin-1-interacting protein 4, dJ468O1.1, EC 6.3.2, EC 6.3.2.-, Itch, itchy (mouse homolog) E3 ubiquitin protein ligase, itchy E3 ubiquitin protein ligase homolog (mouse), itchy homolog E3 ubiquitin protein ligase, NAPP1E3 ubiquitin-protein ligase Itchy homolog, NFE2-associated polypeptide 1, ubiquitin protein ligase ITCH | |
| ITCH antibody was raised against a 15 amino acid peptide near the amino terminus of human ITCH . The immunogen is located within amino acids 40 - 90 of ITCH. Amino Acid Squence: NNTNSPKWKQPLTV | |
| 0.1 mL | |
| Apoptosis, Cell Biology, Immunology, Signal Transduction | |
| 83737 | |
| Store at 4C in the dark. | |
| IgG |
| Western Blot, ELISA | |
| mFluor Violet 500 SE | |
| 50mM Sodium Borate | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction