missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ITCH/AIP4 Antibody (01), DyLight 594, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-06616DL594
This item is not returnable.
View return policy
Description
ITCH/AIP4 Monoclonal antibody specifically detects ITCH/AIP4 in Human samples. It is validated for Flow Cytometry, Immunofluorescence
Specifications
| ITCH/AIP4 | |
| Monoclonal | |
| DyLight 594 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry, Immunofluorescence | |
| 01 | |
| Flow Cytometry, Immunocytochemistry/Immunofluorescence | |
| AIF4, AIP4dJ468O1.1 (atrophin 1 interacting protein 4 (AIP4)), atrophin-1 interacting protein 4, Atrophin-1-interacting protein 4, dJ468O1.1, EC 6.3.2, EC 6.3.2.-, Itch, itchy (mouse homolog) E3 ubiquitin protein ligase, itchy E3 ubiquitin protein ligase homolog (mouse), itchy homolog E3 ubiquitin protein ligase, NAPP1E3 ubiquitin-protein ligase Itchy homolog, NFE2-associated polypeptide 1, ubiquitin protein ligase ITCH | |
| This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human ITCH/AIP4 (NP_113671.3; Arg526-Glu903). | |
| 0.1 mL | |
| Apoptosis, Cell Biology, Immunology, Signal Transduction | |
| 83737 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction