missing translation for 'onlineSavingsMsg'
Learn More

Itaconic Acid, TRC

Product Code. 30402301
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30402301 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30402301 Supplier Toronto Research Chemicals Supplier No. TRCI93100025G
 Restricted Item
Estimated Shipment: 19-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Itaconic Acid
CAS 97-65-4
Molecular Formula C5H6O4
Quantity 25 g
Synonym 2-Methylenebutanedioic Acid, Methylenesuccinic Acid, 2-Methylenebutanedioic Acid, 2-Methylenesuccinic Acid, 2-Propene-1,2-dicarboxylic Acid, 4-Hydroxy-2-methylene-4-oxobutanoic Acid, Methylenebutanedioic Acid, NSC 3357, Propylenedicarboxylic Acid
InChI Formula InChI=1S/C5H6O4/c1-3(5(8)9)2-4(6)7/h1-2H2,(H,6,7)(H,8,9)
SMILES OC(=O)CC(=C)C(=O)O
Molecular Weight (g/mol) 130.1
Formula Weight 130.0266
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.