missing translation for 'onlineSavingsMsg'
Learn More

Itaconic Acid-13C5,d4, TRC

Product Code. 30391922
Change view
Click to view available options
Quantity:
0.25 mg
1 mg
2.5 mg
Unit Size:
0.25mg
1mg
2.5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30391922 2.5 mg 2.5mg
30394500 1 mg 1mg
30408490 0.25 mg 0.25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30391922 Supplier Toronto Research Chemicals Supplier No. TRCI9310022.5MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Tekniske data

Chemical Name or Material Itaconic Acid-13C5,d4
Molecular Formula 13C5H2D4O4
Quantity 2.5 mg
Synonym 2-Methylenebutanedioic Acid-13C5,d4, 2-Methylenesuccinic Acid-13C5,d4, 2-Propene-1,2-dicarboxylic Acid-13C5,d4, 4-Hydroxy-2-methylene-4-oxobutanoic Acid-13C5,d4, Methylenebutanedioic Acid-13C5,d4, Methylenesuccinic Acid-13C5,d4, NSC 3357-13C5,d4, Propylenedicarboxylic Acid-13C5,d4
InChI Formula InChI=1S/C5H6O4/c1-3(5(8)9)2-4(6)7/h1-2H2,(H,6,7)(H,8,9)/i1+1D2,2+1D2,3+1,4+1,5+1
SMILES O[13C](/[13C]([13C]([2H])([2H])[13C](O)=O)=[13C]([2H])/[2H])=O
Molecular Weight (g/mol) 139.09
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.