missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ IT1t dihydrochloride
GREENER_CHOICE

Product Code. 30001566
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30001566 10 mg 10mg
30001589 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30001566 Supplier Tocris Bioscience™ Supplier No. 4596/10
  • ONLINE EXCLUSIVE PRICE
    £208.00
    £196.35 / 10mg
    Save £11.65
    5% Off
 Restricted Item
Estimated Shipment: 03-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Potent CXCR4 antagonist

Potent CXCR4 antagonist (IC50 = 1.1 nM in calcium mobilization assays). Orally available. Blocks interaction with the HIV envelope protein, gp120 (IC50 = 7 nM, inhibition of X4-tropic HIV-1IIIB attachment).
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1092776-63-0
Molecular Formula C21H36Cl2N4S2

Specifications

Quantity 10 mg
Formula Weight 479.57
Purity >99%
Molecular Weight (g/mol) 479.567
Chemical Name or Material IT1t dihydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.