missing translation for 'onlineSavingsMsg'
Learn More

IT1t dihydrochloride, MedChemExpress

Product Code. p-200098481
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16492736

Brand: Medchem Express HY101458A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress IT1t dihydrochloride is a potent CXCR4 antagonist; inhibits CXCL12/CXCR4 interaction with an IC50 of 2.1 nM.

IT1t dihydrochloride is a potent CXCR4 antagonist

Specifications

Chemical Name or Material IT1t dihydrochloride
Physical Form Solid
CAS 1092776-63-0
Color White
Quantity 25 mg
Purity Grade Notes Research
Molecular Formula C21H36Cl2N4S2
For Use With (Application) COVID-19-anti-virus
Solubility Information H2O : 50 mg/mL (104.26 mM; Need ultrasonic) ∣DMSO : ≥ 30 mg/mL (62.56 mM)
SMILES CC1(C)CN2C(CS/C(NC3CCCCC3)=N\C4CCCCC4)=CSC2=N1.[H]Cl.[H]Cl
Molecular Weight (g/mol) 479.57
Formula Weight 479.57
Percent Purity 99.89%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.