missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ISYNA1 Polyclonal Antibody
GREENER_CHOICE

Product Code. 15526352
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15526352 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15526352 Supplier Invitrogen™ Supplier No. PA544105

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Peptide sequence: LQEQLWPHME ALRPRPSVYI PEFIAANQSA RADNLIPGSR AQQLEQIRRD Sequence homology: Cow: 86%; Guinea Pig: 93%; Horse: 86%; Human: 100%; Mouse: 86%; Rat: 86%; Yeast: 92%.

Myoinositol, the most common naturally occurring form of inositol, is a component of plasma membrane phospholipids and functions as a cell signaling molecule. ISYNA1 (EC 5.5.1.4), or IPS, is a rate-limiting enzyme that catalyzes the de novo synthesis of myoinositol 1-phosphate from glucose 6-phosphate. Myoinositol, the most common naturally occurring form of inositol, is a component of plasma membrane phospholipids and functions as a cell signaling molecule. ISYNA1 (EC 5.5.1.4), or IPS, is a rate-limiting enzyme that catalyzes the de novo synthesis of myoinositol 1-phosphate from glucose 6-phosphate (Seelan et al., 2004 ).
TRUSTED_SUSTAINABILITY

Specifications

Antigen ISYNA1
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene ISYNA1
Gene Accession No. Q9NPH2
Gene Alias 1300017C10Rik; AU018670; hINO1; Ino1; INOS; Inositol-3-phosphate synthase 1; IPS; IPS 1; IPS-1; Isyna1; MGC133800 protein; MI-1-P synthase; MIP synthase; Myo-inositol 1-phosphate synthase; Myo-inositol 1-phosphate synthase A1; myo-inositol-1-phosphate synthase; testis secretory sperm-binding protein Li 200a
Gene Symbols ISYNA1
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal of human ISYNA1 (aa 161-210).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51477
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.