missing translation for 'onlineSavingsMsg'
Learn More

ISYNA1 Polyclonal Antibody
GREENER_CHOICE

Product Code. 16676754
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16676754 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16676754 Supplier Invitrogen Supplier No. PA5100272

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

ISYNA1 Polyclonal Antibody

ISYNA1 Polyclonal Antibody for Western Blot

TRUSTED_SUSTAINABILITY

Specifications

Antigen ISYNA1
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene ISYNA1
Gene Accession No. Q6AYK3, Q9JHU9, Q9NPH2
Gene Alias 1300017C10Rik; AU018670; hINO1; Ino1; INOS; Inositol-3-phosphate synthase 1; IPS; IPS 1; IPS-1; Isyna1; MGC133800 protein; MI-1-P synthase; MIP synthase; Myo-inositol 1-phosphate synthase; Myo-inositol 1-phosphate synthase A1; myo-inositol-1-phosphate synthase; testis secretory sperm-binding protein Li 200a
Gene Symbols ISYNA1
Host Species Rabbit
Immunogen A synthesized peptide derived from human ISYNA1(Accession Q9NPH2), corresponding to amino acid residues L515-T558.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 290651, 51477, 71780
Target Species Human, Mouse, Rat, Monkey
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.