missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ISX 9, Tocris Bioscience™
Neurogenic agent; induces neuronal differentiation of SVZ progenitors and also induces cardiomyogenic differentiation
£181.49 - £727.08
Chemical Identifiers
CAS | 832115-62-5 |
---|---|
Molecular Formula | C11H10N2O2S |
Molecular Weight (g/mol) | 234.273 |
InChI Key | SYENTKHGMVKGAQ-UHFFFAOYSA-N |
PubChem CID | 19582717 |
IUPAC Name | N-cyclopropyl-5-thiophen-2-yl-1,2-oxazole-3-carboxamide |
SMILES | C1CC1NC(=O)C2=NOC(=C2)C3=CC=CS3 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18044022
|
Tocris Bioscience
4439/10 |
10 mg |
£181.49
10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18034242
|
Tocris Bioscience
4439/50 |
50 mg |
£727.08
50mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Neurogenic agent. Mediates neuroD reporter gene induction via activation of Ca2+ influx; increases expression of neurogenic differentiation 1 (NeuroD1) transcription factor. Induces neuronal differentiation in human cortical neuronal cells (HCN), adult mouse whole brain (MWB) and subventricular zone (SVZ) progenitors. Also shown to induce cardiomyogenic differentiation of Notch-activated epicardium-derived cells in vitro.Chemical Identifiers
832115-62-5 | |
234.273 | |
19582717 | |
C1CC1NC(=O)C2=NOC(=C2)C3=CC=CS3 |
C11H10N2O2S | |
SYENTKHGMVKGAQ-UHFFFAOYSA-N | |
N-cyclopropyl-5-thiophen-2-yl-1,2-oxazole-3-carboxamide |
Specifications
832115-62-5 | |
isx-9, isx 9, n-cyclopropyl-5-thiophen-2-yl isoxazole-3-carboxamide, isoxazole 9 isx-9, neuronal differentiation inducer iii, n-cyclopropyl-5-thiophen-2-yl-1,2-oxazole-3-carboxamide, isoxazole 9, isx9, isoxazole 9 isx9, n-cyclopropyl-5-2-thienyl-3-isoxazolecarboxamide | |
C1CC1NC(=O)C2=NOC(=C2)C3=CC=CS3 | |
234.273 | |
234.27 | |
ISX 9 |
C11H10N2O2S | |
SYENTKHGMVKGAQ-UHFFFAOYSA-N | |
N-cyclopropyl-5-thiophen-2-yl-1,2-oxazole-3-carboxamide | |
19582717 | |
>99% |
Safety and Handling
Recommended Storage : Store at +4°C