Learn More
STEMCELL Technologies ISX-9
Inducer of neural differentiation
Brand:  STEMCELL Technologies 73202
Encompass_Preferred
Product Code. 17190182
  • £191.70 / 10mg
Description
  • ISX-9 is a small molecule inducer of adult neural stem cell differentiation both in vitro and in vivo (Schneider et al.) It has been shown to act through a calcium-activated signaling pathway dependent on myocyte-enhancer factor 2 (MEF2)-dependent gene expression (Schneider et al.; Petrik et al.)