missing translation for 'onlineSavingsMsg'
Learn More

Istradefylline, MedChemExpress

Product Code. 16450577
Encompass_Preferred
Click to view available options
Quantity:
10 mg
50 mg
100 mg
200 mg
500 mg
Unit Size:
100mg
10mg
200mg
500mg
50mg
This item is not returnable. View return policy

Product Code. 16450577

Brand: Medchem Express HY10888

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Istradefylline is a very potent, selective and orally active adenosine A2A receptor antagonist with Ki of 2.2 nM in experimental models of Parkinson's disease.

Istradefylline is a very potent, selective and orally active adenosine A2A receptor antagonist with Ki of 2.2 nM in experimental models of Parkinson's disease.

Specifications

Chemical Name or Material Istradefylline
CAS 155270-99-8
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C20H24N4O4
Quantity 50 mg
Synonym KW-6002
Solubility Information DMSO : 25.33 mg/mL (65.89 mM; Need ultrasonic and warming)
SMILES O=C1C2=C(N=C(/C=C/C3=CC(OC)=C(OC)C=C3)N2C)N(CC)C(N1CC)=O
Molecular Weight (g/mol) 384.43
Formula Weight 384.43
Percent Purity 98.62%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neurodegeneration
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.