missing translation for 'onlineSavingsMsg'
Learn More

ISRIB, TRC

Product Code. 30391544
Change view
Click to view available options
Quantity:
100 mg
250 mg
Unit Size:
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30391544 250 mg 250mg
30405037 100 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30391544 Supplier Toronto Research Chemicals Supplier No. TRCI926000250MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1597403-47-8
Molecular Formula C22H24Cl2N2O4
Molecular Weight (g/mol) 451.34
Synonym N,N'-trans-1,4-Cyclohexanediylbis[2-(4-chlorophenoxy)acetamide], SMDC 750213
SMILES Clc1ccc(OCC(=O)N[C@@H]2CC[C@H](CC2)NC(=O)COc3ccc(Cl)cc3)cc1

Specifications

Quantity 250 mg
InChI Formula InChI=1S/C22H24Cl2N2O4/c23-15-1-9-19(10-2-15)29-13-21(27)25-17-5-7-18(8-6-17)26-22(28)14-30-20-11-3-16(24)4-12-20/h1-4,9-12,17-18H,5-8,13-14H2,(H,25,27)(H,26,28)
Formula Weight 450.1113
Percent Purity >95%
Chemical Name or Material ISRIB
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.