missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ISPD Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA565803
This item is not returnable.
View return policy
Description
Immunogen sequence: IVVAVTGENME VMKSIIQKYQ HKRISLVEAG VTRHRSIFNG LKALAEDQIN SKLSKPEVVI IHDAVRPFVE EGVLLKVVTA AKEHG Highest antigen sequence identity to the following orthologs - mouse 83%, rat 80%.
Required for protein O-linked mannosylation. Probably acts as a nucleotidyltransferase involved in synthesis of a nucleotide sugar. Required for dystroglycan O-mannosylation.
Specifications
| ISPD | |
| Polyclonal | |
| Unconjugated | |
| ISPD | |
| 2-C-methyl-D-erythritol 4-phosphate cytidylyltransferase-like protein; 4930579E17Rik; 4-diphosphocytidyl-2C-methyl-D-erythritol synthase homolog; AV040780; Crppa; D-ribitol-5-phosphate cytidylyltransferase; hCG_1745121; hISPD; isoprenoid synthase domain containing; isoprenoid synthase domain-containing protein; Ispd; MDDGA7; MDDGC7; Nip; notch1-induced protein; testicular tissue protein Li 97 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 729920 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry | |
| 0.4 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| A4D126 | |
| ISPD | |
| Recombinant Human ISPD. Recombinant protein control fragment (Product #RP-106475). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction