missing translation for 'onlineSavingsMsg'
Learn More

Isoxicam, TRC

Product Code. 30134534
Change view
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30134534 10 g 10g
17991445 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30134534 Supplier Toronto Research Chemicals Supplier No. TRCI91814510G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 34552-84-6
Molecular Formula C14 H13 N3 O5 S
Molecular Weight (g/mol) 335.34
IUPAC Name 4-hydroxy-2-methyl-N-(5-methyl-1,2-oxazol-3-yl)-1,1-dioxo-1λ^{6},2-benzothiazine-3-carboxamide
SMILES CN1C(=C(O)c2ccccc2S1(=O)=O)C(=O)Nc3cc(C)on3

Specifications

Quantity 10 g
InChI Formula InChI=1S/C14H13N3O5S/c1-8-7-11(16-22-8)15-14(19)12-13(18)9-5-3-4-6-10(9)23(20,21)17(12)2/h3-7,18H,1-2H3,(H,15,16,19)
Formula Weight 335.0576
Chemical Name or Material Isoxicam
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.