missing translation for 'onlineSavingsMsg'
Learn More

Isoxazole-5-carboxylic acid, 98%, Thermo Scientific™

Product Code. 10630883 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
Packaging:
Glass bottle
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10630883 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10630883 Supplier Thermo Scientific Acros Supplier No. 355480010

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 21169-71-1
Melting Point 137°C to 138°C
Color Yellow
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Molecular Formula C4H3NO3
MDL Number MFCD00156151
Quantity 1 g
Synonym isoxazole-5-carboxylic acid, 5-isoxazolecarboxylic acid, isoxazole-5-carboxylicacid, 5-carboxyisoxazole, pubchem8643, isoxazole-5-carboxylic, 5-carboxy-1,2-oxazole, isoxazole 5-carboxylic acid, ksc207s2h
InChI Key MIIQJAUWHSUTIT-UHFFFAOYSA-N
SMILES C1=C(ON=C1)C(=O)O
IUPAC Name 1,2-oxazole-5-carboxylic acid
Molecular Weight (g/mol) 113.07
PubChem CID 2060599
Formula Weight 113.07
Percent Purity 98%
Physical Form Powder
Chemical Name or Material Isoxazole-5-carboxylic acid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.