missing translation for 'onlineSavingsMsg'
Learn More

Isoxazole-5-carbothioamide, 97%, Thermo Scientific™

Catalog Number 10100612
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
175334-72-2
C4H4N2OS
128.15
MFCD00276995
CEDRGDFENMZKCQ-UHFFFAOYSA-N
isoxazole-5-carbothioamide, 5-isoxazolecarbothioamide, isoxazole-5-carbothioic acid amide, 5-isoxazolecarbothioamide 9ci, aminoisoxazol-5-ylmethane-1-thione
2799515
1,2-oxazole-5-carbothioamide
NC(=S)C1=CC=NO1
This item is not returnable. View return policy
175334-72-2
C4H4N2OS
128.15
MFCD00276995
CEDRGDFENMZKCQ-UHFFFAOYSA-N
isoxazole-5-carbothioamide, 5-isoxazolecarbothioamide, isoxazole-5-carbothioic acid amide, 5-isoxazolecarbothioamide 9ci, aminoisoxazol-5-ylmethane-1-thione
2799515
1,2-oxazole-5-carbothioamide
NC(=S)C1=CC=NO1
CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 128.15
Percent Purity 97%
Chemical Name or Material Isoxazole-5-carbothioamide
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'