missing translation for 'onlineSavingsMsg'
Learn More

Isoxazole-5-carbonyl chloride, 97%

Product Code. 11447576
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11447576 5 g 5g
11437576 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11447576 Supplier Thermo Scientific Alfa Aesar Supplier No. A14625.06
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 62348-13-4 | C4H2ClNO2 | 131.52 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 62348-13-4
Molecular Formula C4H2ClNO2
Molecular Weight (g/mol) 131.52
MDL Number MFCD00067868
InChI Key NASLINFISOTVJJ-UHFFFAOYSA-N
Synonym isoxazole-5-carbonyl chloride, 5-isoxazolecarbonyl chloride, isoxazole-5-carbonylchloride, isoxazole-5-carboxylic acid chloride, acmc-1bhwq, 5-isoxazolecarbonylchloride, isoxazol-5-carbonyl chloride, isoxazole-5-carbonyl-chloride, 5-isoxazolecarbonyl chloride 9ci
PubChem CID 2736707
IUPAC Name 1,2-oxazole-5-carbonyl chloride
SMILES ClC(=O)C1=CC=NO1

Specifications

Density 1.362
Boiling Point 167°C to 168°C
Refractive Index 1.499
Quantity 5 g
UN Number UN3265
Beilstein 4375023
Sensitivity Moisture sensitive
Formula Weight 131.52
Percent Purity 97%
Chemical Name or Material Isoxazole-5-carbonyl chloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.