missing translation for 'onlineSavingsMsg'
Learn More

Isoxathion, TRC

Product Code. 30401870
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
1mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30401870 10 mg -
30391583 1 mg 1mg
30402774 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30401870 Supplier Toronto Research Chemicals Supplier No. TRCI91769010MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 18854-01-8
Molecular Formula C13 H16 N O4 P S
Molecular Weight (g/mol) 313.31
Synonym Phosphorothioic acid, O,O-diethyl O-(5-phenyl-3-isoxazolyl) ester, 3-Isoxazolol, 5-phenyl-, O-ester with O,O-diethyl phosphorothioate (8CI), Isoxathion, Karphos
IUPAC Name diethoxy-[(5-phenyl-1,2-oxazol-3-yl)oxy]-sulfanylidene-λ^{5}-phosphane
SMILES CCOP(=S)(OCC)Oc1cc(on1)c2ccccc2

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C13H16NO4PS/c1-3-15-19(20,16-4-2)18-13-10-12(17-14-13)11-8-6-5-7-9-11/h5-10H,3-4H2,1-2H3
Formula Weight 313.0538
Chemical Name or Material Isoxathion
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.