missing translation for 'onlineSavingsMsg'
Learn More

Isoxathion, TRC

Product Code. 30402774
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
1mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30402774 5 mg 5mg
30391583 1 mg 1mg
30401870 10 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30402774 Supplier Toronto Research Chemicals Supplier No. TRCI9176905MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 18854-01-8
Molecular Formula C13 H16 N O4 P S
Molecular Weight (g/mol) 313.31
Synonym Phosphorothioic acid, O,O-diethyl O-(5-phenyl-3-isoxazolyl) ester, 3-Isoxazolol, 5-phenyl-, O-ester with O,O-diethyl phosphorothioate (8CI), Isoxathion, Karphos
IUPAC Name diethoxy-[(5-phenyl-1,2-oxazol-3-yl)oxy]-sulfanylidene-λ^{5}-phosphane
SMILES CCOP(=S)(OCC)Oc1cc(on1)c2ccccc2

Specifications

Quantity 5 mg
InChI Formula InChI=1S/C13H16NO4PS/c1-3-15-19(20,16-4-2)18-13-10-12(17-14-13)11-8-6-5-7-9-11/h5-10H,3-4H2,1-2H3
Formula Weight 313.0538
Chemical Name or Material Isoxathion
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.