missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ ISOX DUAL
Dual BRD4 and CBP/p300 bromodomain inhibitor
Specifications
| Chemical Name or Material | [3-[4-[2-[5-(Dimethyl-1,2-oxazol-4-yl)-1-[2-(morpholin-4-yl)ethyl]-1H-1,3-benzodiazol-2-yl]ethyl]phenoxy]propyl]dimethylamine |
|---|---|
| CAS | 1962928-22-8 |
Description
Dual CBP/p300 and BRD4 bromodomain inhibitor (IC50 values are 0.65 and 1.5 μM for CBP and BRD4 respectively). Downregulates IL-6, IL-1β and IFN-β in macrophages.Specifications
| [3-[4-[2-[5-(Dimethyl-1,2-oxazol-4-yl)-1-[2-(morpholin-4-yl)ethyl]-1H-1,3-benzodiazol-2-yl]ethyl]phenoxy]propyl]dimethylamine | |
| 1962928-22-8 |
Safety and Handling
Recommended Storage : Store at 4°C
Spot an opportunity for improvement?
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.