missing translation for 'onlineSavingsMsg'
Learn More

Isowyosine, TRC

Product Code. 30396076
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30396076 50 mg 50mg
30401869 100 mg -
30397910 10 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30396076 Supplier Toronto Research Chemicals Supplier No. TRCI91863550MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 577773-09-2
Molecular Formula C14H17N5O5
Molecular Weight (g/mol) 335.32
Synonym 3-((2R,3R,4S,5R)-3,4-Dihydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)-6,7-dimethyl-3H-imidazo[1,2-a]purin-9(4H)-one, 3,4-Dihydro-6,7-dimethyl-3-beta-D-ribofuranosyl-9H-imidazo[1,2-a]purin-9-one, imG2
IUPAC Name 3-[(2R,3R,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]-6,7-dimethyl-4H-imidazo[1,2-a]purin-9-one
SMILES Cc1nc2Nc3c(ncn3[C@@H]4O[C@H](CO)[C@@H](O)[C@H]4O)C(=O)n2c1C

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C14H17N5O5/c1-5-6(2)19-12(23)8-11(17-14(19)16-5)18(4-15-8)13-10(22)9(21)7(3-20)24-13/h4,7,9-10,13,20-22H,3H2,1-2H3,(H,16,17)/t7-,9-,10-,13-/m1/s1
Formula Weight 335.123
Chemical Name or Material Isowyosine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.