missing translation for 'onlineSavingsMsg'
Learn More

Isovitexin, MedChemExpress

Product Code. 16499088
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
Unit Size:
10mg
25mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16499088 10 mg 10mg
16493896 25 mg 25mg
16401508 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16499088 Supplier Medchem Express Supplier No. HYN0773

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Isovitexin is a flavonoid isolated from rice hulls of Oryza sativa, possesses anti-inflammatory and anti-oxidant activities; Isovitexin acts like a JNK1/2 inhibitor and inhibits the activation of NF-κB.

Isovitexin is a flavonoid isolated from rice hulls of Oryza sativa, possesses anti-inflammatory and anti-oxidant activities

Specifications

Chemical Name or Material Isovitexin
CAS 38953-85-4
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C21H20O10
Quantity 10 mg
Synonym Saponaretin Homovitexin
Solubility Information DMSO : 25 mg/mL (57.82 mM; Need ultrasonic)
SMILES OC1=C(C2=O)C(OC(C3=CC=C(O)C=C3)=C2)=CC(O)=C1[C@@H]([C@@H]([C@@H](O)[C@@H]4O)O)O[C@@H]4CO
Molecular Weight (g/mol) 432.38
Formula Weight 432.38
Percent Purity 97.98%
Grade Research
Physical Form Powder
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.