missing translation for 'onlineSavingsMsg'
Learn More

Isovaleryl chloride, 97%

Product Code. 11486896
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11486896 25 g 25g
11496896 100 g 100g
11406906 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11486896 Supplier Thermo Scientific Alfa Aesar Supplier No. B24002.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 108-12-3 | C5H9ClO | 120.576 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 108-12-3
Molecular Formula C5H9ClO
Molecular Weight (g/mol) 120.576
MDL Number MFCD00000738
InChI Key ISULZYQDGYXDFW-UHFFFAOYSA-N
Synonym isovaleryl chloride, butanoyl chloride, 3-methyl, isovaleroyl chloride, isopentanoyl chloride, isovaleric acid chloride, 3-methylbutyryl chloride, isovalerylchloride, 3-methyl-butanoyl chloride, unii-j2ml32m57v, 3-methyl-butyryl chloride
PubChem CID 66054
IUPAC Name 3-methylbutanoyl chloride
SMILES CC(C)CC(=O)Cl

Specifications

Density 0.989
Boiling Point 115°C to 117°C
Flash Point 27°C (80°F)
Refractive Index 1.416
Quantity 25 g
UN Number UN2920
Beilstein 741910
Sensitivity Moisture sensitive
Merck Index 14,5232
Formula Weight 120.58
Percent Purity 98%
Chemical Name or Material Isovaleryl chloride
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.