missing translation for 'onlineSavingsMsg'
Learn More

Isovaleric Acid Ethyl-d5 Ester, TRC

Product Code. 30395777
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30395777 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30395777 Supplier Toronto Research Chemicals Supplier No. TRCI917577250MG
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Isovaleric Acid Ethyl-d5 Ester
CAS 1082581-99-4
Molecular Formula C7H9D5O2
Quantity 250 mg
Synonym 3-Methylbutanoic Acid Ethyl-d5 Ester, 3-Methylbutyric Acid Ethyl-d5 Ester, Ethyl-d5 3-Methylbutanoate, Ethyl-d5 3-Methylbutyrate, Ethyl-d5 Isopentanoate, Ethyl-d5 Isovalerate, Isopentanoic Acid Ethyl-d5 Ester, NSC 8869-d5
InChI Formula InChI=1S/C7H14O2/c1-4-9-7(8)5-6(2)3/h6H,4-5H2,1-3H3/i1D3,4D2
SMILES O=C(CC(C)C)OC([2H])([2H])C([2H])([2H])[2H]
Molecular Weight (g/mol) 135.22
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.