missing translation for 'onlineSavingsMsg'
Learn More

Isovaleric acid-benzyl ester, Dr. Ehrenstorfer

Product Code. 30326932
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
This item is not returnable. View return policy

Product Code. 30326932

Brand: Dr. Ehrenstorfer DREC14479480

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Isovaleric acid-benzyl ester
CAS 103-38-8
Analyte or Component Names Isovaleric acid-benzyl ester
Shipping Condition Room Temperature
Molecular Formula C12 H16 O2
Certifications/Compliance ISO/IEC 17025
Synonym Isovaleric acid, benzyl ester (6CI,8CI), Benzyl 3-methylbutanoate, Benzyl 3-methylbutyrate, Benzyl isopentanoate, Benzyl isovalerate, NSC 46124, Phenylmethyl isovalerate
InChI Formula InChI=1S/C12H16O2/c1-10(2)8-12(13)14-9-11-6-4-3-5-7-11/h3-7,10H,8-9H2,1-2H3
Solution Type Neat
SMILES CC(C)CC(=O)OCc1ccccc1
IUPAC Name benzyl 3-methylbutanoate
Industry Type Environmental Testing
Molecular Weight (g/mol) 192.25
Quantity 250 mg
Formula Weight 192.115 g/mol
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.