missing translation for 'onlineSavingsMsg'
Learn More

Isovaleric acid, 99%

Product Code. 10398833 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 g
100 g
500 g
2.5 kg
Packaging:
Glass bottle
Unit Size:
100g
10g
2.5kg
500g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10398833 2.5 kg Glass bottle 2.5kg
10226060 10 g Glass bottle 10g
10153540 100 g Glass bottle 100g
10041730 500 g Glass bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Sea freight only: For EU stock allow up to 15 days delivery.
Product Code. 10398833 Supplier Thermo Scientific Acros Supplier No. 156690025
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Sea freight only: For EU stock allow up to 15 days delivery.

CAS: 503-74-2 | C5H10O2 | 102.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 503-74-2
Molecular Formula C5H10O2
Molecular Weight (g/mol) 102.13
MDL Number MFCD00002726
InChI Key GWYFCOCPABKNJV-UHFFFAOYSA-N
Synonym isovaleric acid, isopentanoic acid, 3-methylbutyric acid, delphinic acid, isopropylacetic acid, butanoic acid, 3-methyl, isovalerianic acid, isobutylformic acid, 3-methylbutyrate, beta-methylbutyric acid
PubChem CID 10430
ChEBI CHEBI:28484
IUPAC Name 3-methylbutanoic acid
SMILES CC(C)CC(O)=O

Specifications

Melting Point -35.0°C
Color Colorless to Yellow
Density 0.9260g/mL
Boiling Point 176.0°C
Flash Point 70°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula (CH3)2CHCH2CO2H
Refractive Index 1.3990 to 1.4070
Quantity 2.5 kg
Beilstein 02, 309
Merck Index 15, 5277
Specific Gravity 0.926
Solubility Information Solubility in water: 25g/L (20°C). Other solubilities: soluble in alcohol,chloroform and ether
Viscosity 2.4 mPa.s (20°C)
Formula Weight 102.13
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Isovaleric acid, 99%
Show More Show Less
compliance-icons
Product Identifier
  • Isovaleric acid
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.