missing translation for 'onlineSavingsMsg'
Learn More

Isotretinoin, MedChemExpress

Product Code. 16476636
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16476636 100 mg 100mg
16441756 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16476636 Supplier Medchem Express Supplier No. HY15127
 Restricted Item
Estimated Shipment: 11-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Isotretinoin(13-cis-Retinoic acid) is a medication used for the treatment of severe acne.

Isotretinoin(13-cis-Retinoic acid) is a medication used for the treatment of severe acne.

Specifications

Chemical Name or Material Isotretinoin
CAS 4759-48-2
Color Orange
Purity Grade Notes Research
Molecular Formula C20H28O2
Quantity 100 mg
Synonym 13-cis-Retinoic acid
Solubility Information DMSO : 50 mg/mL (166.42 mM; Need ultrasonic)
SMILES CC1(C)C(/C=C/C(C)=C/C=C/C(C)=C\C(O)=O)=C(C)CCC1
Molecular Weight (g/mol) 300.44
Formula Weight 300.44
Percent Purity 99.53%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.