missing translation for 'onlineSavingsMsg'
Learn More

Isotretinoin, MedChemExpress

Product Code. 16441756
Encompass_Preferred
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
This item is not returnable. View return policy

Product Code. 16441756

Brand: Medchem Express HY15127

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Isotretinoin(13-cis-Retinoic acid) is a medication used for the treatment of severe acne.

Isotretinoin(13-cis-Retinoic acid) is a medication used for the treatment of severe acne.

Specifications

Chemical Name or Material Isotretinoin
CAS 4759-48-2
Color Orange
Purity Grade Notes Research
Molecular Formula C20H28O2
Quantity 500 mg
Synonym 13-cis-Retinoic acid
Solubility Information DMSO : 50 mg/mL (166.42 mM; Need ultrasonic)
SMILES CC1(C)C(/C=C/C(C)=C/C=C/C(C)=C\C(O)=O)=C(C)CCC1
Molecular Weight (g/mol) 300.44
Formula Weight 300.44
Percent Purity 99.53%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.