missing translation for 'onlineSavingsMsg'
Learn More

Isotrazodone, TRC

Product Code. 30401780
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30401780 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30401780 Supplier Toronto Research Chemicals Supplier No. TRCI91751510MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Isotrazodone
CAS 157072-18-9
Molecular Formula C19 H22 Cl N5 O
Quantity 10 mg
Synonym 1-[3-[4-(3-chlorophenyl)-1-piperazinyl]propyl]-3-oxo-1,2,4-Triazolo[4,3-a]pyridinium, 1-[3-[4-(3-chlorophenyl)-1-piperazinyl]propyl]-2,3-dihydro-3-oxo-1,2,4-Triazolo[4,3-a]pyridinium (9CI), 1-{3-[4-(3-Chlorophenyl)piperazin-1-yl]propyl}-[1,2,4]triazolo[4,3-a]pyridin-1-ium-3-olate, 1-{3-[4-(3-Chlorophenyl)piperazin-1-yl]propyl}-[1,2,4]triazolo[4,3-a]pyridin-1-ium-3-olate
InChI Formula InChI=1S/C19H22ClN5O/c20-16-5-3-6-17(15-16)23-13-11-22(12-14-23)8-4-10-25-18-7-1-2-9-24(18)19(26)21-25/h1-3,5-7,9,15H,4,8,10-14H2
SMILES [O-]c1n[n+](CCCN2CCN(CC2)c3cccc(Cl)c3)c4ccccn14
IUPAC Name 1-[3-[4-(3-chlorophenyl)piperazin-1-yl]propyl]-[1,2,4]triazolo[4,3-a]pyridin-1-ium-3-olate
Molecular Weight (g/mol) 371.86
Formula Weight 371.1513
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.