missing translation for 'onlineSavingsMsg'
Learn More

Beckman Coulter™ ISOTON™ II Diluent

Product Code. 12754878 Shop All Beckman Coulter Life Sciences Products
Encompass
Change view
Click to view available options
Quantity:
20 L
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
12754878 20 L Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12754878 Supplier Beckman Coulter™ Supplier No. 844 80 11

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Beckman Coulter™ ISOTON™ II Diluent is a filtered, phosphate-buffered saline solution compatible with human blood cells, and may be used for the suspension of most biological cells and many industrial samples.

Beckman Coulter™ ISOTON™ II Diluent is a necessary reagent for the daily maintenance and operation of Beckman Coulter flow cytometers, including Multisizer particle counter appliances. The Multisizer is a highly versatile particle counting and characterization system. It uses the Coulter principle to detect particles via electrical zone sensing, regardless of the nature of the particle or its optical properties. This makes it an ideal tool for detecting and counting a wide variety of particles.

Specifications

Content And Storage 2°C to 30°C
For Use With (Equipment) Flow cytometers, including Multisizer particle counter appliances
pH 7.35-7.65
Quantity 20 L
Form Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.