missing translation for 'onlineSavingsMsg'
Learn More

Isothiazole, 97%, Thermo Scientific Chemicals

Product Code. 15529573 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15529573 1 g 1g
15539573 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15529573 Supplier Thermo Scientific Acros Supplier No. 458420010
 Restricted Item
Estimated Shipment: 12-11-2026
Add to basket
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 288-16-4
Molecular Formula C3H3NS
Molecular Weight (g/mol) 85.13
InChI Key ZLTPDFXIESTBQG-UHFFFAOYSA-N
Synonym isothiazole, 2-azathiophene, 1,2-thiazol, isothiazole;1,2-thiazole, 4-fluoro-3'-methylbenzophenone
PubChem CID 67515
ChEBI CHEBI:35600
IUPAC Name 1,2-thiazole
SMILES C1=CSN=C1

Specifications

Flash Point 22°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 85.13
Percent Purity 97%
Chemical Name or Material Isothiazole
compliance-icons compliance-icons
Product Identifier
  • Isothiazole
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P264-Wash face, hands and any thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.