missing translation for 'onlineSavingsMsg'
Få mere at vide

Isothiazol-3(2H)-one, TRC

Artikelnummer. 30390145
Skift visning
Klik for at se tilgængelige muligheder
Quantity:
50 mg
Pakningsstørrelse:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Quantity Pakningsstørrelse
30390145 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 30390145 Leverandør Toronto Research Chemicals Leverandørnr. TRCI91762050MG
 Restricted Item
Estimated Shipment: 12-11-2026
Læg i indkøbskurv
Læg i indkøbskurv

missing translation for 'validMainframeMsg'

Denne vare kan ikke returneres. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Isothiazol-3(2H)-one
CAS 1003-07-2
Molecular Formula C3 H3 N O S
Quantity 50 mg
Synonym 4-Isothiazolin-3-one (7CI,8CI), 1,2-Isothiazole-3-one, 1,2-Thiazol-3-one, 3-Hydroxyisothiazole, 3-Isothiazolol, 3-Isothiazolone, 3-Oxo-2,3-dihydroisothiazole, A 01, Duo Mei Jing, Ebotec aqua, GT 2060, GY 506, Isothiazolin-3-one, Isothiazolinone, NSC 93489, PT 490, Xarcin AB
InChI Formula InChI=1S/C3H3NOS/c5-3-1-2-6-4-3/h1-2H,(H,4,5)
SMILES O=C1NSC=C1
IUPAC Name 1,2-thiazol-3-one
Molecular Weight (g/mol) 101.127
Formula Weight 100.9935
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.