missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Isosorbide Mononitrate, Mikromol™
Description
Isosorbide Mononitrate
Specifications
Specifications
| Chemical Name or Material | Isosorbide Mononitrate |
| CAS | 16051-77-7 |
| Analyte or Component Names | Isosorbide Mononitrate |
| Shipping Condition | Room Temperature |
| Molecular Formula | C6 H9 N O6 |
| Certifications/Compliance | ISO 17034 |
| Synonym | Isosorbide Dinitrate, Diluted Imp. C (EP), Isosorbide Mononitrate, Isosorbide Dinitrate Imp. C (EP), Isosorbide 5-Nitrate, 1,4:3,6-Dianhydro-D-glucitol 5-nitrate, D-1,4:3,6-Dianhydro-D-glucitol 5-nitrate, Furo[3,2-b]furan, D-glucitol deriv., AHR 4698, BM 22-145, Conpin, Conpin Retardkaps, Corangin, Corangin SR, Dilavenil, Duride, Elan, Elantan, Elantan Long, Elantan Retard, Geomatrix, IS 5MN, ISMN, Imdur, Imdur 60, Imdur Durules, Imodur, Imtrate, Ismexin, Ismo, Ismo 20, Ismox, Isomon, Isomonat, Isomonit, Isopen 20, Isosorbide 5-mononitrate, Isosorbide 5-nitrate, Isosorbide mononitrate, Iturol, Medocor, Momo Mack, Monicor, Monis, Monit, Monit 20, Monizid, Mono Corax, Mono Corax Retard, Mono-Cedocard, Mono-Mack, Mono-Sanorania, Monoclair, Monocord 20, Monocord 40, Monocord 50SR, Monodur Durules, Monoket, Monoket OD, Monoket Retard, Monolong, Monolong 40, Monolong 60, Mononit, Mononit 20, Mononit 40, Mononit Retard 50, Monopront, Monosorb, Monosorbitrate, Monosordil, Monotrate, Monovas, Multitab, Nitramin, Olicard, Ormox, Pentacard, Pentacard 20, Vasotrate, Isosorbide Dinitrate Impurity C |
| InChI Formula | InChI=1S/C6H9NO6/c8-3-1-11-6-4(13-7(9)10)2-12-5(3)6/h3-6,8H,1-2H2/t3-,4+,5+,6+/m0/s1 |
| SMILES | O[C@H]1CO[C@@H]2[C@@H](CO[C@H]12)O[N+](=O)[O-] |
| IUPAC Name | [(3S,3aR,6R,6aS)-3-hydroxy-2,3,3a,5,6,6a-hexahydrofuro[3,2-b]furan-6-yl] nitrate |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?