missing translation for 'onlineSavingsMsg'
Learn More

Isosorbide dimethyl ether, 98%

Product Code. 11408137
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11408137 25 g 25g
11468710 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11408137 Supplier Thermo Scientific Alfa Aesar Supplier No. A17516.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 5306-85-4 | C8H14O4 | 174.20 g/mol

It is employed as a specialty solvent. It is a solvent for active ingredients especially DHA for sunless tanning products. It is also utilized as viscosity controlling agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed as a specialty solvent. It is a solvent for active ingredients especially DHA for sunless tanning products. It is also utilized as viscosity controlling agent.

Solubility
Soluble in water.

Notes
Store in a cool place. Ensure proper ventilation. Keep away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5306-85-4
Molecular Formula C8H14O4
Molecular Weight (g/mol) 174.20
MDL Number MFCD00064828
InChI Key MEJYDZQQVZJMPP-UHFFFAOYNA-N
Synonym dimethyl isosorbide, isosorbide dimethyl ether, 3r,3ar,6s,6ar-3,6-dimethoxyhexahydrofuro 3,2-b furan, unii-sa6a6v432s, 1,4:3,6-dianhydro-2,5-di-o-methyl-d-glucitol, arlasolve dmi, 2,5-di-o-methyl-1,4:3,6-dianhydro-d-glucitol, dimethyl isosorbide ether, 3r,3ar,6s,6ar-3,6-dimethoxy-hexahydrofuro 3,2-b furan, 3s,3ar,6r,6ar-3,6-dimethoxy-2,3,3a,5,6,6a-hexahydrofuro 3,2-b furan
PubChem CID 62990
IUPAC Name (3S,3aR,6R,6aR)-3,6-dimethoxy-2,3,3a,5,6,6a-hexahydrofuro[3,2-b]furan
SMILES COC1COC2C(COC12)OC

Specifications

Melting Point -70°C
Density 1.165
Quantity 25 g
Refractive Index 1.462
Beilstein 80854
Boiling Point 93°C to 95°C (0.1 mmHg)
Solubility Information Soluble in water.
Formula Weight 174.2
Percent Purity 98%
Chemical Name or Material Isosorbide dimethyl ether

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.