missing translation for 'onlineSavingsMsg'
Learn More

Isorhynchophylline, MedChemExpress

Product Code. 16471498
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16471498 5 mg 5mg
16459088 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16471498 Supplier Medchem Express Supplier No. HYN0766
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Isorhynchophylline (IRN), an alkaloid isolated from Uncaria rhynchophylla, possesses the effects of lowered blood pressure, vasodilatation and protection against ischemia-induced neuronal damage.

Isorhynchophylline (IRN), an alkaloid isolated from Uncaria rhynchophylla, possesses the effects of lowered blood pressure, vasodilatation and protection against ischemia-induced neuronal damage.

Specifications

Chemical Name or Material Isorhynchophylline
CAS 1-4-6859
Color White
Purity Grade Notes Research
Molecular Formula C22H28N2O4
Quantity 5 mg
Solubility Information DMSO : 100 mg/mL (260.10 mM; Need ultrasonic)
SMILES O=C(NC1=C2C=CC=C1)[C@@]32[C@@](C[C@H](/C(C(OC)=O)=C\OC)[C@@H](CC)C4)([H])N4CC3
Molecular Weight (g/mol) 384.47
Formula Weight 384.47
Percent Purity 98.21%
Grade Research
Physical Form Solid
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.