missing translation for 'onlineSavingsMsg'
Learn More

Isoquinoline, TRC

Product Code. 30403506
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30403506 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30403506 Supplier Toronto Research Chemicals Supplier No. TRCI87493010G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Isochinolin
CAS 119-65-3
Molecular Formula C9 H7 N
Quantity 10 g
Synonym Isoquinoline (6CI, 8CI, 9CI, ACI), 2-Azanaphthalene, 2-Benzazine, Benzo[c]pyridine, Benzopyridine, NSC 3395, β-Quinoline
InChI Formula InChI=1S/C9H7N/c1-2-4-9-7-10-6-5-8(9)3-1/h1-7H
SMILES c1ccc2cnccc2c1
IUPAC Name isoquinoline
Molecular Weight (g/mol) 129.16
Formula Weight 129.0578
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.