missing translation for 'onlineSavingsMsg'
Learn More

Isoquinoline, 97%

Product Code. 11479177
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11479177 500 g 500g
11469177 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11479177 Supplier Thermo Scientific Alfa Aesar Supplier No. B21279.36
 Restricted Item
Estimated Shipment: 31-07-2026
Add to basket
This item is not returnable. View return policy

CAS: 119-65-3 | C9H7N | 129.16 g/mol

Isoquinoline is used in the manufacture of dyes, paints, antifungal. It is also used as a solvent for the extraction of resins and terpenes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Isoquinoline is used in the manufacture of dyes, paints, antifungal. It is also used as a solvent for the extraction of resins and terpenes.

Solubility
Soluble in water (5g/L).

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 119-65-3
Molecular Formula C9H7N
Molecular Weight (g/mol) 129.16
MDL Number MFCD00006898,MFCD31699977
InChI Key AWJUIBRHMBBTKR-UHFFFAOYSA-N
Synonym 2-azanaphthalene, 2-benzazine, isochinolin, benzo c pyridine, beta-quinoline, 2-benzanine, 3,4-benzopyridine, .beta.-quinoline, isochinolin czech
PubChem CID 8405
ChEBI CHEBI:16092
IUPAC Name isoquinoline
SMILES C1=CC=C2C=NC=CC2=C1

Specifications

Melting Point 24°C to 27°C
Density 1.09
pH 7.5
Boiling Point 241°C to 243°C
Flash Point 107°C (225°F)
Odor Pungent
Refractive Index 1.6245
Quantity 500 g
UN Number UN2811
Beilstein 107549
Merck Index 14,5222
Solubility Information Soluble in water (5g/L).
Formula Weight 129.16
Percent Purity ≥97%
Chemical Name or Material Isoquinoline
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.