missing translation for 'onlineSavingsMsg'
Learn More

Isoquinoline-5-sulfonyl Chloride, Hydrochloride, TRC

Product Code. 30388845
Change view
Click to view available options
Quantity:
500 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30388845 500 mg
30408608 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30388845 Supplier Toronto Research Chemicals Supplier No. TRCI878000500MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 105627-79-0
Molecular Formula C9 H6 Cl N O2 S . Cl H
Molecular Weight (g/mol) 264.13
IUPAC Name isoquinoline-5-sulfonyl chloride;hydrochloride
SMILES Cl.ClS(=O)(=O)c1cccc2cnccc12

Specifications

Quantity 500 mg
InChI Formula InChI=1S/C9H6ClNO2S.ClH/c10-14(12,13)9-3-1-2-7-6-11-5-4-8(7)9;/h1-6H;1H
Formula Weight 262.9575
Chemical Name or Material Isoquinoline-5-sulfonyl Chloride Hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.