missing translation for 'onlineSavingsMsg'
Learn More

Isoquinoline-5-carboxylic acid, 96%

Product Code. 11335435
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
Product Code. Quantity unitSize
11335435 250 mg 250mg
11325435 1 g 1g
2 options
This item is not returnable. View return policy

Product Code. 11335435

Brand: Thermo Scientific Alfa Aesar L19637.MD

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 27810-64-6 | C10H7NO2 | 173.171 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 27810-64-6
Molecular Formula C10H7NO2
Molecular Weight (g/mol) 173.171
MDL Number MFCD03788744
InChI Key ZIPLFLRGHZAXSJ-UHFFFAOYSA-N
Synonym 5-isoquinolinecarboxylic acid, 5-carboxyisoquinoline, isoquinoline-5-carboxylicacid, pubchem19508, acmc-1ch46, isoquinoline-5-carboxylic acid
PubChem CID 260936
IUPAC Name isoquinoline-5-carboxylic acid
SMILES C1=CC2=C(C=CN=C2)C(=C1)C(=O)O

Specifications

Melting Point 278°C to 281°C
Boiling Point 60°C to 62°C (11 mmHg)
Quantity 250 mg
Beilstein 129180
Formula Weight 173.17
Percent Purity 96%
Chemical Name or Material Isoquinoline-5-carboxylic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.