missing translation for 'onlineSavingsMsg'
Learn More

Isoquinoline-5-boronic acid, 97%

Product Code. 15479987
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15479987 250 mg 250mg
15469987 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15479987 Supplier Thermo Scientific Alfa Aesar Supplier No. H53230.MD
 Restricted Item
Estimated Shipment: 30-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 371766-08-4 | C9H8BNO2 | 172.978 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 371766-08-4
Molecular Formula C9H8BNO2
Molecular Weight (g/mol) 172.978
MDL Number MFCD03839356
InChI Key XKEYHBLSCGBBGU-UHFFFAOYSA-N
Synonym isoquinoline-5-boronic acid, 5-isoquinolineboronic acid, isoquinolin-5-yl-5-boronic acid, boronic acid, 5-isoquinolinyl, isoquinolin-5-yl boronic acid, 5-isoquinolineboronicacid, isoquinoline-5-boric acid, 5-boronoisoquinoline, acmc-209ird, 5-isoquinolylboronic acid
PubChem CID 599474
IUPAC Name isoquinolin-5-ylboronic acid
SMILES B(C1=C2C=CN=CC2=CC=C1)(O)O

Specifications

Melting Point 204°C to 206°C
Quantity 250 mg
Formula Weight 172.98
Percent Purity 97%
Chemical Name or Material Isoquinoline-5-boronic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.