missing translation for 'onlineSavingsMsg'
Learn More

Isoquinoline-4-boronic acid 2,2-dimethylpropanediol-1,3 cyclic ester, 97%, Thermo Scientific™

Product Code. 10621085
Change view
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10621085 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10621085 Supplier Thermo Scientific Maybridge Supplier No. CC04940DA
 Restricted Item
Estimated Shipment: 26-10-2026
Add to basket
This item is not returnable. View return policy

Specifications

CAS 844891-01-6
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Molecular Formula C14H16BNO2
MDL Number MFCD06659058
Quantity 1 g
Synonym 4-5,5-dimethyl-1,3,2-dioxaborinan-2-yl isoquinoline, isoquinoline-4-boronic acid 2,2-dimethylpropanediol-1,3 cyclic ester, isoquinoline-4-boronic acid, 2,2-dimethylpropane-1,3-diol cyclic ester, isoquinoline-4-boronic acid neopentyl glycol ester, 3 cyclic ester, akos brn-0509, 2-dimethylpropanediol-1, isoquinoline-4-boronic acid 2, 2-4-isoquinolyl-5,5-dimethyl-1,3,2-dioxaborane, isoquinoline,4-5,5-dimethyl-1,3,2-dioxaborinan-2-yl
InChI Key AQEZCUCPPAKCLQ-UHFFFAOYSA-N
SMILES CC1(C)COB(OC1)C1=CN=CC2=CC=CC=C12
IUPAC Name 4-(5,5-dimethyl-1,3,2-dioxaborinan-2-yl)isoquinoline
Molecular Weight (g/mol) 241.10
PubChem CID 2795158
Formula Weight 241.1
Percent Purity 97%
Chemical Name or Material Isoquinoline-4-boronic acid 2, 2-dimethylpropanediol-1, 3 cyclic ester
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.