missing translation for 'onlineSavingsMsg'
Learn More

Isoquinolin-3-amine, 97%, Thermo Scientific™

Product Code. 10130242
Change view
Click to view available options
Quantity:
250 mg
1 g
10 g
Packaging:
Amber Glass Bottle
Unit Size:
10g
1g
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10130242 1 g Amber Glass Bottle 1g
10039091 250 mg Amber Glass Bottle 250mg
10214644 10 g Amber Glass Bottle 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10130242 Supplier Thermo Scientific Maybridge Supplier No. BTB10019DA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 25475-67-6
Molecular Formula C9H8N2
Molecular Weight (g/mol) 144.177
InChI Key VYCKDIRCVDCQAE-UHFFFAOYSA-N
Synonym 3-aminoisoquinoline, 3-isoquinolinamine, isoquinolin-3-ylamine, isoquinoline, 3-amino, 3-isoquinolylamine, isoquinoline-3-amine, chembl61743, 2h-isoquinolin-3-imine, isoquinolin-3-imine, zlchem 309
PubChem CID 311869
IUPAC Name isoquinolin-3-amine
SMILES C1=CC=C2C=NC(=CC2=C1)N

Specifications

Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 144.18
Percent Purity 97%
Chemical Name or Material Isoquinolin-3-amine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.