missing translation for 'onlineSavingsMsg'
Learn More

Isopulegol, TRC

Product Code. 30389815
Change view
Click to view available options
Quantity:
1 g
2.5 g
5 g
Unit Size:
1g
2.5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30389815 5 g -
30407865 2.5 g 2.5g
30393373 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30389815 Supplier Toronto Research Chemicals Supplier No. TRCI8747405G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7786-67-6
Molecular Formula C10H18O
Molecular Weight (g/mol) 154.25
Synonym 5-Methyl-2-(1-methylethenyl)cyclohexanol, 2-Isopropenyl-5-methylcyclohexanol, 5-Methyl-2-(1-methylvinyl)cyclohexanol, p-Menth-8-en-3-ol
IUPAC Name 5-methyl-2-prop-1-en-2-ylcyclohexan-1-ol
SMILES CC1CCC(C(O)C1)C(=C)C

Specifications

Quantity 5 g
InChI Formula InChI=1S/C10H18O/c1-7(2)9-5-4-8(3)6-10(9)11/h8-11H,1,4-6H2,2-3H3
Formula Weight 154.1358
Chemical Name or Material Isopulegol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.